(2-Bromophenyl)(3,5-dimethyl-1H-pyrazol-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

166494-1G 166494-500MG 166494-5G
77465-542EA 826.06 USD
77465-542 77478-300 77488-480
(2-Bromophenyl)(3,5-dimethyl-1H-pyrazol-1-yl)methanone ≥97%
(2-Bromophenyl)(3,5-dimethyl-1H-pyrazol-1-yl)methanone

Precaución: For research use only

Formula: C₁₂H₁₁BrN₂O
MW: 279.1325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00585032
CAS Number: 414879-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR