3-(2,5-Dichlorophenyl)-3-oxetanol ≥97%

Supplier: AOBChem USA

Warning

166364-1G 166364-500MG 166364-5G 166364-25G
77465-534EA 284.28 USD
77465-534 77478-292 77488-472 77499-096
3-(2,5-Dichlorophenyl)-3-oxetanol ≥97%
3-(2,5-Dichlorophenyl)-3-oxetanol

Precaución: For research use only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34851909
CAS Number: 1531857-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR