3-[4-(Ethylthio)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166357-1G 166357-500MG 166357-5G
77465-530EA 541.78 USD
77465-530 77478-288 77488-468
3-[4-(Ethylthio)phenyl]-3-oxetanol ≥95%
3-[4-(Ethylthio)phenyl]-3-oxetanol

Precaución: For research use only

Formula: C₁₁H₁₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD34851902
CAS Number: 1889496-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR