3-[4-(2-Methylpropyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166347-1G 166347-500MG 166347-5G 166347-25G
77465-526EA 393.46 USD
77465-526 77478-284 77488-464 77499-092
3-[4-(2-Methylpropyl)phenyl]-3-oxetanol ≥95%
3-[4-(2-Methylpropyl)phenyl]-3-oxetanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851893
CAS Number: 1895392-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR