3-(4-Bromo-3-chloro-2-fluorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166335-1G 166335-500MG 166335-5G
77465-522EA 646.84 USD
77465-522 77478-280 77488-460
3-(4-Bromo-3-chloro-2-fluorophenyl)-3-oxetanol ≥95%
3-(4-Bromo-3-chloro-2-fluorophenyl)-3-oxetanol

Precaución: For research use only

Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34851881
CAS Number: 2103908-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR