3-(2,3-Difluorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166305-1G 166305-500MG 166305-5G
77465-518EA 522.21 USD
77465-518 77478-276 77488-456
3-(2,3-Difluorophenyl)-3-oxetanol ≥95%
3-(2,3-Difluorophenyl)-3-oxetanol

Precaución: For research use only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851851
CAS Number: 2111218-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR