3-(3-Hydroxy-3-oxetanyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

166283-1G 166283-500MG 166283-5G
77465-514EA 895.07 USD
77465-514 77478-272 77488-452
3-(3-Hydroxy-3-oxetanyl)benzonitrile ≥95%
3-(3-Hydroxy-3-oxetanyl)benzonitrile

Precaución: For research use only

Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851831
CAS Number: 2733483-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR