3-(1-Naphthalenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166271-1G 166271-500MG 166271-5G
77465-510EA 434.66 USD
77465-510 77478-268 77488-448
3-(1-Naphthalenyl)-3-oxetanol ≥95%
3-(1-Naphthalenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₃H₁₂O₂
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851821
CAS Number: 1888958-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR