3-(3-Methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166260-1G 166260-500MG 166260-5G
77465-506EA 433.63 USD
77465-506 77478-264 77488-444
3-(3-Methylphenyl)-3-oxetanol ≥95%
3-(3-Methylphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34646509
CAS Number: 1511615-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR