3-(2,4-Dimethoxyphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166250-1G 166250-500MG 166250-5G
77465-502EA 541.78 USD
77465-502 77478-260 77488-440
3-(2,4-Dimethoxyphenyl)-3-oxetanol ≥95%
3-(2,4-Dimethoxyphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34851803
CAS Number: 1401867-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR