3-(2-Bromo-6-fluorophenyl)oxetan-3-ol ≥97%

Supplier: AOBChem USA

Warning

166217-1G 166217-500MG 166217-5G 166217-25G
77465-498EA 370.8 USD
77465-498 77478-256 77488-436 77499-088
3-(2-Bromo-6-fluorophenyl)oxetan-3-ol ≥97%
3-(2-Bromo-6-fluorophenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₉H₈BrFO₂
MW: 247.0610 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851795
CAS Number: 2969427-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR