3-(2-Fluoro-5-methoxyphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

166211-1G 166211-500MG 166211-5G 166211-25G
77465-494EA 415.09 USD
77465-494 77478-252 77488-432 77499-086
3-(2-Fluoro-5-methoxyphenyl)oxetan-3-ol ≥95%
3-(2-Fluoro-5-methoxyphenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34851793
CAS Number: 2969433-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR