3-(3-Bromo-2,6-difluorophenyl)oxetan-3-ol ≥97%

Supplier: AOBChem USA

Warning

166205-1G 166205-500MG 166205-5G
77465-490EA 646.84 USD
77465-490 77478-248 77488-428
3-(3-Bromo-2,6-difluorophenyl)oxetan-3-ol ≥97%
3-(3-Bromo-2,6-difluorophenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₉H₇BrF₂O₂
MW: 265.0514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851791
CAS Number: 2103968-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR