3-(4-Methoxyphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

166199-1G 166199-500MG 166199-5G 166199-25G
77465-486EA 296.64 USD
77465-486 77478-244 77488-424 77499-082
3-(4-Methoxyphenyl)oxetan-3-ol ≥95%
3-(4-Methoxyphenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD27664934
CAS Number: 26755-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR