3-(3-Bromophenyl)oxetan-3-ol ≥97%

Supplier: AOBChem USA

Warning

166187-1G 166187-500MG 166187-5G 166187-25G
77465-482EA 417.15 USD
77465-482 77478-240 77488-420 77499-080
3-(3-Bromophenyl)oxetan-3-ol ≥97%
3-(3-Bromophenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22370048
CAS Number: 1379811-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR