(2,6-Difluoro-3-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165938-1G 165938-500MG 165938-5G 165938-25G
77465-478EA 415.09 USD
77465-478 77478-236 77488-416 77499-076
(2,6-Difluoro-3-methylphenyl)(ethyl)sulfane ≥95%
(2,6-Difluoro-3-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂S
MW: 188.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851613
CAS Number: 2921849-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR