(2,3-Dichloro-6-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165935-1G 165935-500MG 165935-5G 165935-25G
77465-474EA 415.09 USD
77465-474 77478-232 77488-412 77499-072
(2,3-Dichloro-6-fluorophenyl)(ethyl)sulfane ≥95%
(2,3-Dichloro-6-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇Cl₂FS
Storage Temperature: Ambient
MDL Number: MFCD34851610
CAS Number: 2921852-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR