(6-Chloro-2-fluoro-3-methoxyphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165925-1G 165925-500MG 165925-5G 165925-25G
77465-470EA 415.09 USD
77465-470 77478-228 77488-408 77499-068
(6-Chloro-2-fluoro-3-methoxyphenyl)(ethyl)sulfane ≥95%
(6-Chloro-2-fluoro-3-methoxyphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀ClFOS
MW: 220.692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851602
CAS Number: 2921849-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR