(2-Bromo-6-fluoro-4-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165915-1G 165915-500MG 165915-5G 165915-25G
77465-466EA 415.09 USD
77465-466 77478-224 77488-404 77499-064
(2-Bromo-6-fluoro-4-methylphenyl)(ethyl)sulfane ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFS
Storage Temperature: Ambient
MDL Number: MFCD34851592
CAS Number: 2921897-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR