(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165912-1G 165912-500MG 165912-5G 165912-25G
77465-462EA 415.09 USD
77465-462 77478-220 77488-400 77499-060
(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(ethyl)sulfane ≥95%
(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₅H₁₄BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34851589
CAS Number: 2921742-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR