(2-Bromo-3-chloro-6-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165909-1G 165909-500MG 165909-5G 165909-25G
77465-458EA 415.09 USD
77465-458 77478-216 77488-396 77499-056
(2-Bromo-3-chloro-6-fluorophenyl)(ethyl)sulfane ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇BrClFS
Storage Temperature: Ambient
MDL Number: MFCD34851586
CAS Number: 2921836-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR