(2,6-Difluoro-4-methoxyphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165871-1G 165871-500MG 165871-5G 165871-25G
77465-450EA 415.09 USD
77465-450 77478-210 77488-388 77499-048
(2,6-Difluoro-4-methoxyphenyl)(ethyl)sulfane ≥95%
(2,6-Difluoro-4-methoxyphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂OS
MW: 204.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851553
CAS Number: 2921860-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR