2-(Ethylthio)-5-(mEthylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

165853-1G 165853-500MG 165853-5G 165853-25G
77465-446EA 415.09 USD
77465-446 77478-204 77488-382 77499-044
2-(Ethylthio)-5-(mEthylthio)thiophene ≥95%
2-(Ethylthio)-5-(mEthylthio)thiophene

Precaución: For research use only

Formula: C₇H₁₀S₃
Storage Temperature: Ambient
MDL Number: MFCD34851536
CAS Number: 61809-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR