(E)-1-(2,3-dichloro-6-fluorophenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165842-1G 165842-500MG 165842-5G 165842-25G
77465-442EA 415.09 USD
77465-442 77478-202 77488-380 77499-040
(E)-1-(2,3-dichloro-6-fluorophenyl)but-2-en-1-ol ≥95%
(E)-1-(2,3-dichloro-6-fluorophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₉Cl₂FO
MW: 235.082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851526
CAS Number: 2921913-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR