(E)-1-(3,6-dibromo-2-fluorophenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165821-1G 165821-500MG 165821-5G 165821-25G
77465-430EA 415.09 USD
77465-430 77478-190 77488-368 77499-028
(E)-1-(3,6-dibromo-2-fluorophenyl)but-2-en-1-ol ≥95%
(E)-1-(3,6-dibromo-2-fluorophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₉Br₂FO
MW: 323.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851506
CAS Number: 2921913-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR