(E)-1-(6-Bromo-2-fluoro-3-methylphenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165815-1G 165815-500MG 165815-5G 165815-25G
77465-422EA 415.09 USD
77465-422 77478-182 77488-360 77499-020
(E)-1-(6-Bromo-2-fluoro-3-methylphenyl)but-2-en-1-ol ≥95%
(E)-1-(6-Bromo-2-fluoro-3-methylphenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34851500
CAS Number: 2921913-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR