(E)-1-(5-Chloro-2-fluoro-3-methylphenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165757-1G 165757-500MG 165757-5G 165757-25G
77465-414EA 415.09 USD
77465-414 77478-174 77488-352 77499-014
(E)-1-(5-Chloro-2-fluoro-3-methylphenyl)but-2-en-1-ol ≥95%
(E)-1-(5-Chloro-2-fluoro-3-methylphenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD34851445
CAS Number: 2921913-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR