1,3-Dibromo-2-(3-bromopropoxy)benzene ≥96%

Supplier: AOBChem USA

Warning

165198-1G 165198-500MG 165198-5G 165198-25G
77465-410EA 233.81 USD
77465-410 77478-170 77488-346 77499-008
1,3-Dibromo-2-(3-bromopropoxy)benzene ≥96%
1,3-Dibromo-2-(3-bromopropoxy)benzene

Precaución: For research use only

Formula: C₉H₉Br₃O
Storage Temperature: Ambient
MDL Number: MFCD11164558
CAS Number: 206347-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR