1-(cyclohexyloxy)-4-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

164977-1G 164977-500MG 164977-5G 164977-25G
77465-406EA 246.17 USD
77465-406 77478-166 77488-342 77499-004
1-(cyclohexyloxy)-4-iodobenzene ≥95%
1-(cyclohexyloxy)-4-iodobenzene

Precaución: For research use only

Formula: C₁₂H₁₅IO
MW: 302.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18360943
CAS Number: 851199-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR