2-(2-Bromo-3-chloro-6-fluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

16488-1G 16488-500MG 16488-5G 16488-25G
77465-402EA 363.59 USD
77465-402 77478-162 77488-338 77499-000
2-(2-Bromo-3-chloro-6-fluorophenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-3-chloro-6-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976877
CAS Number: 2221812-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR