2-Bromo-1-fluoro-3-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

164802-1G 164802-500MG 164802-5G 164802-25G
77465-398EA 415.09 USD
77465-398 77478-158 77488-334 77498-996
2-Bromo-1-fluoro-3-phenoxybenzene ≥97%
2-Bromo-1-fluoro-3-phenoxybenzene

Precaución: For research use only

Formula: C₁₂H₈BrFO
MW: 267.094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850865
CAS Number: 1800321-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR