3-Bromo-5-iodo-6-methylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

164679-1G 164679-500MG 164679-5G 164679-25G
77465-394EA 369.77 USD
77465-394 77478-154 77488-330 77498-994
3-Bromo-5-iodo-6-methylpyridin-2-amine ≥97%
3-Bromo-5-iodo-6-methylpyridin-2-amine

Precaución: For research use only

Formula: C₆H₆BrIN₂
MW: 312.9337 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07138516
CAS Number: 875001-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR