3-(2-Fluoro-5-methylphenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164666-1G 164666-500MG 164666-5G 164666-25G
77465-390EA 415.09 USD
77465-390 77478-150 77488-326 77498-992
3-(2-Fluoro-5-methylphenyl)pentan-3-ol ≥97%
3-(2-Fluoro-5-methylphenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.2611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222308
CAS Number: 1340401-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR