3-(2-Fluoro-5-methoxyphenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164664-1G 164664-500MG 164664-5G 164664-25G
77465-386EA 415.09 USD
77465-386 77478-146 77488-322 77498-990
3-(2-Fluoro-5-methoxyphenyl)pentan-3-ol ≥97%
3-(2-Fluoro-5-methoxyphenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34850799
CAS Number: 2969470-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR