3-(3-(trifluoromethyl)phenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164662-1G 164662-500MG 164662-5G 164662-25G
77465-382EA 415.09 USD
77465-382 77478-142 77488-318 77498-986
3-(3-(trifluoromethyl)phenyl)pentan-3-ol ≥97%
3-(3-(trifluoromethyl)phenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₅F₃O
MW: 232.2421 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222299
CAS Number: 76481-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR