3-(4-Bromophenyl)pentan-3-ol ≥96%

Supplier: AOBChem USA

Warning

164659-1G 164659-500MG 164659-5G 164659-25G
77465-378EA 297.67 USD
77465-378 77478-138 77488-314 77498-982
3-(4-Bromophenyl)pentan-3-ol ≥96%
3-(4-Bromophenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.1402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27929839
CAS Number: 850918-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR