2-(3-Bromo-2,6-difluorophenyl)butan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164641-1G 164641-500MG 164641-5G
77465-374EA 447.02 USD
77465-374 77478-134 77488-308
2-(3-Bromo-2,6-difluorophenyl)butan-2-ol ≥97%
2-(3-Bromo-2,6-difluorophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34850797
CAS Number: 1700518-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR