2-(4-Chloro-2-fluorophenyl)butan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164637-1G 164637-500MG 164637-5G 164637-25G
77465-370EA 397.58 USD
77465-370 77478-130 77488-304 77498-976
2-(4-Chloro-2-fluorophenyl)butan-2-ol ≥97%
2-(4-Chloro-2-fluorophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD23852787
CAS Number: 1537324-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR