2-(2-Fluoro-5-methylphenyl)butan-2-ol ≥96%

Supplier: AOBChem USA

Warning

164633-1G 164633-500MG 164633-5G 164633-25G
77465-366EA 415.09 USD
77465-366 77478-126 77488-300 77498-972
2-(2-Fluoro-5-methylphenyl)butan-2-ol ≥96%
2-(2-Fluoro-5-methylphenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.2346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222292
CAS Number: 1341142-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR