2-(4-Bromophenyl)butan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164624-1G 164624-500MG 164624-5G 164624-25G
77465-362EA 229.69 USD
77465-362 77478-122 77488-296 77498-968
2-(4-Bromophenyl)butan-2-ol ≥97%
2-(4-Bromophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₃BrO
MW: 229.1136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00268089
CAS Number: 58977-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR