1-(4-Bromophenyl)pentan-1-ol ≥97%

Supplier: AOBChem USA

Warning

164584-1G 164584-500MG 164584-5G
77465-358EA 471.74 USD
77465-358 77478-118 77488-290
1-(4-Bromophenyl)pentan-1-ol ≥97%
1-(4-Bromophenyl)pentan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.1402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786807
CAS Number: 76287-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR