1-(2-Bromo-6-fluorophenyl)butan-1-ol ≥97%

Supplier: AOBChem USA

Warning

164580-1G 164580-500MG 164580-5G 164580-25G
77465-354EA 226.6 USD
77465-354 77478-114 77488-286 77498-962
1-(2-Bromo-6-fluorophenyl)butan-1-ol ≥97%
1-(2-Bromo-6-fluorophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.1040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26136421
CAS Number: 1602786-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR