1-(3-Bromo-2-fluorophenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

164578-1G 164578-500MG 164578-5G 164578-25G
77465-350EA 397.58 USD
77465-350 77478-110 77488-282 77498-958
1-(3-Bromo-2-fluorophenyl)butan-1-ol ≥95%
1-(3-Bromo-2-fluorophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.1040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850787
CAS Number: 1699655-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR