1-(4-(trifluoromethyl)phenyl)butan-1-ol ≥97%

Supplier: AOBChem USA

Warning

164575-1G 164575-500MG 164575-5G
77465-346EA 471.74 USD
77465-346 77478-106 77488-278
1-(4-(trifluoromethyl)phenyl)butan-1-ol ≥97%
1-(4-(trifluoromethyl)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD16310044
CAS Number: 115840-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR