(3-Bromophenyl)(butyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164566-1G 164566-500MG 164566-5G 164566-25G
77465-342EA 86.52 USD
77465-342 77478-102 77488-274 77498-952
(3-Bromophenyl)(butyl)sulfane ≥97%
(3-Bromophenyl)(butyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃BrS
MW: 245.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20040716
CAS Number: 156021-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR