Butyl(phenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

164562-1G 164562-500MG 164562-5G 164562-25G
77465-338EA 124.63 USD
77465-338 77478-100 77488-270 77498-948
Butyl(phenyl)sulfane ≥95%
Butyl(phenyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.2831 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040696
CAS Number: 1126-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR