(2-Bromo-6-fluorophenyl)(propyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164560-1G 164560-500MG 164560-5G 164560-25G
77465-334EA 415.09 USD
77465-334 77478-096 77488-266 77498-944
(2-Bromo-6-fluorophenyl)(propyl)sulfane ≥97%
(2-Bromo-6-fluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFS
Storage Temperature: Ambient
MDL Number: MFCD34850782
CAS Number: 2921735-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR