(2,5-Dichlorophenyl)(propyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164553-1G 164553-500MG 164553-5G 164553-25G
77465-326EA 159.65 USD
77465-326 77478-088 77488-256 77498-940
(2,5-Dichlorophenyl)(propyl)sulfane ≥97%
(2,5-Dichlorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀Cl₂S
Storage Temperature: Ambient
MDL Number: MFCD26792639
CAS Number: 16155-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR