(3-Bromophenyl)(propyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164549-1G 164549-500MG 164549-5G 164549-25G
77465-322EA 133.9 USD
77465-322 77478-084 77488-252 77498-934
(3-Bromophenyl)(propyl)sulfane ≥97%
(3-Bromophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁BrS
Storage Temperature: Ambient
MDL Number: MFCD20040719
CAS Number: 883443-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR