(4-Chlorophenyl)(propyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164545-1G 164545-500MG 164545-5G 164545-25G
77465-314EA 172.01 USD
77465-314 77478-076 77488-242 77498-928
(4-Chlorophenyl)(propyl)sulfane ≥97%
(4-Chlorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD26792616
CAS Number: 16155-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR